missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-pyridinecarboxylic Acid, TRC

Codice prodotto. 30482457
Cambia vista
Click to view available options
Quantity:
25 g
Dimensione della confezione:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
30482457 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30482457 Fornitore Toronto Research Chemicals N. del fornitore TRCM27167025G
 Restricted Item
Estimated Shipment: 10-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methoxy-2-pyridinecarboxylic Acid
CAS 29082-91-5
Molecular Formula C7H7NO3
Quantity 25 g
Synonym 4-Methoxypicolinic Acid
InChI Formula InChI=1S/C7H7NO3/c1-11-5-2-3-8-6(4-5)7(9)10/h2-4H,1H3,(H,9,10)
SMILES O=C(O)C1=NC=CC(OC)=C1
IUPAC Name 4-Methoxy-2-pyridinecarboxylic Acid
Molecular Weight (g/mol) 153.14
Formula Weight 153.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.