missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-nitrobenzonitrile, 97%, Thermo Scientific™

Product Code. 10041164
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 38469-83-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 137°C to 140°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H6N2O3
Linear Formula CNC6H3(NO2)OCH3
MDL Number MFCD00667741
Quantity 5 g
Synonym 2-nitro-4-methoxybenzonitrile, 4-cyano-3-nitroanisole, 4-methoxy-2-nitro-benzonitrile, benzonitrile, 4-methoxy-2-nitro, pubchem16089, acmc-209xbl, 3-nitro-4-cyanoanisole, ksc580a1d, benzonitrile,4-methoxy-2-nitro, 4-methoxy-2-nitrobenzonitrile
InChI Key UUYPUFCWSUBTFP-UHFFFAOYSA-N
SMILES COC1=CC=C(C#N)C(=C1)[N+]([O-])=O
IUPAC Name 4-methoxy-2-nitrobenzonitrile
Molecular Weight (g/mol) 178.15
PubChem CID 288665
Formula Weight 178.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2-nitrobenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-2-nitrobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'