missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2-nitroaniline, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.
Solubility
Slightly soluble in water. Soluble in hot methanol almost transparency.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.
Solubility
Slightly soluble in water. Soluble in hot methanol almost transparency.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications
Melting Point | 126°C to 128°C |
Quantity | 100 g |
UN Number | UN2811 |
Beilstein | 880318 |
Solubility Information | Slightly soluble in water. Soluble in hot methanol almost transparency. |
Formula Weight | 168.15 |
Percent Purity | 99% |
Chemical Name or Material | 4-Methoxy-2-nitroaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'