missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-nitroaniline(2-Nitro-p-anisidine), TRC

Product Code. 30467648
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30467648 2.5 g 2.5g
30466575 250 mg -
2 options
This item is not returnable. View return policy

Product Code. 30467648

Brand: Toronto Research Chemicals TRCN4918002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-96-8
Formula Weight 168.0535
InChI Formula InChI=1S/C7H8N2O3/c1-12-5-2-3-6(8)7(4-5)9(10)11/h2-4H,8H2,1H3
IUPAC Name 4-methoxy-2-nitroaniline
Molecular Formula C7 H8 N2 O3
Molecular Weight (g/mol) 168.15
SMILES COc1ccc(N)c(c1)[N+](=O)[O-]
Synonym Benzenamine, 4-methoxy-2-nitro-, 4-Methoxy-2-nitroaniline
Chemical Name or Material 2-Nitro-p-anisidine

Specifications

Quantity 2.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.