missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylphenylhydrazine hydrochloride, 96%

Product Code. 11313696
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313696 5 g 5g
11303696 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313696 Supplier Thermo Scientific Alfa Aesar Supplier No. L14810.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93048-16-9 | C8H13ClN2O | 188.655 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93048-16-9
Molecular Formula C8H13ClN2O
Molecular Weight (g/mol) 188.655
MDL Number MFCD03426021
InChI Key OIFKWIPSJGNUCC-UHFFFAOYSA-N
Synonym 4-methoxy-2-methylphenylhydrazine hydrochloride, 4-methoxy-2-methylphenyl hydrazine hydrochloride, 1-4-methoxy-2-methylphenyl hydrazine hydrochloride, 4-hydrazino-3-methylanisole hydrochloride, 4-methoxy-2-methylphenylhydrazinehydrochloride, 4-methoxy-2-methylphenyl-hydrazine hydrochloride, hydrazine, 4-methoxy-2-methylphenyl-, monohydrochloride
PubChem CID 21997741
IUPAC Name (4-methoxy-2-methylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C=CC(=C1)OC)NN.Cl

Specifications

Melting Point ∽110°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 7227704
Formula Weight 188.66
Percent Purity 96%
Chemical Name or Material 4-Methoxy-2-methylphenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.