missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylbenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 11334556
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334556 1 g 1g
11344556 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334556 Supplier Thermo Scientific Alfa Aesar Supplier No. L20112.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Reactant for preparation of biologically active molecule preparation of hydroxyphenylnaphthols as 17ß-hydroxysteroid dehydrogenase Type 2 inhibitors

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208399-66-0
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.98
MDL Number MFCD02684315
InChI Key AMSQNQJCBXQYEX-UHFFFAOYSA-N
Synonym 4-methoxy-2-methylphenyl boronic acid, 2-methyl-4-methoxyphenylboronic acid, 4-methoxy-2-methylbenzeneboronic acid, 4-methoxy-2-methylphenyl boranediol, 4-methoxy-2-methyl-phenyl boronic acid, 4-methoxy-2-methylphenbylboronic acid, 4-methoxy-2-methylphenylboronicacid, boronic acid, 4-methoxy-2-methylphenyl, 2-methyl-4-methoxybenzeneboronic acid, acmc-1cge6
PubChem CID 2773486
IUPAC Name (4-methoxy-2-methylphenyl)boronic acid
SMILES COC1=CC=C(B(O)O)C(C)=C1

Specifications

Melting Point 191°C
Quantity 1 g
Beilstein 8111222
Solubility Information Insoluble in water.
Formula Weight 165.99
Percent Purity 98%
Chemical Name or Material 4-Methoxy-2-methylbenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.