missing translation for 'onlineSavingsMsg'
Learn More

4-Meth-5-(4,4,5,5-tetrameth-1,3,2-dioxaborol-2-yl)-2-(4-(triflmeth)phen)-1,3-thiaz, 97%, Thermo Scientific™

Catalog Number 10370453
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
690631-96-0
C17H19BF3NO2S
369.21
MFCD05664416
ZUROXICAUJQTIB-UHFFFAOYSA-N
4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl thiazole, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-boronic acid pinacol ester, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl ph-1,3-thiazole, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole
2794643
4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
CC1=C(SC(=N1)C1=CC=C(C=C1)C(F)(F)F)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
690631-96-0
C17H19BF3NO2S
369.21
MFCD05664416
ZUROXICAUJQTIB-UHFFFAOYSA-N
4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl thiazole, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-boronic acid pinacol ester, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl ph-1,3-thiazole, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole
2794643
4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
CC1=C(SC(=N1)C1=CC=C(C=C1)C(F)(F)F)B1OC(C)(C)C(C)(C)O1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 369.21
Percent Purity 97%
Chemical Name or Material 4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'