missing translation for 'onlineSavingsMsg'
Learn More

4-Meth-5-(4,4,5,5-tetrameth-1,3,2-dioxaborol-2-yl)-2-(4-(triflmeth)phen)-1,3-thiaz, 97%, Thermo Scientific™

Product Code. 10014292
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10014292 250 mg Amber Glass Bottle 250mg
10370453 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10014292 Supplier Thermo Scientific Maybridge Supplier No. MO00919CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 690631-96-0
Molecular Formula C17H19BF3NO2S
Molecular Weight (g/mol) 369.21
MDL Number MFCD05664416
InChI Key ZUROXICAUJQTIB-UHFFFAOYSA-N
Synonym 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl thiazole, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-boronic acid pinacol ester, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl ph-1,3-thiazole, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole
PubChem CID 2794643
IUPAC Name 4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
SMILES CC1=C(SC(=N1)C1=CC=C(C=C1)C(F)(F)F)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 369.21
Percent Purity 97%
Chemical Name or Material 4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.