missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptopyridine, 96%

Product Code. 10266020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266020 10 g Glass bottle 10g
10717123 1 g Glass bottle 1g
10609843 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10266020 Supplier Thermo Scientific Acros Supplier No. 164710100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4556-23-4 | C5H5NS | 111.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4556-23-4
Molecular Formula C5H5NS
Molecular Weight (g/mol) 111.16
MDL Number MFCD00006422
InChI Key FHTDDANQIMVWKZ-UHFFFAOYSA-N
Synonym 4-mercaptopyridine, pyridine-4-thiol, 4-pyridinethiol, 4-thiopyridine, 4-pyridinethione, pyridine, 4-mercapto, 4-thiopyridone, 4 1h-pyridinethione, pyridine-4 1h-thione, 5-21-07-00167 beilstein handbook reference
PubChem CID 2723889
SMILES S=C1C=CNC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 182°C to 189°C
Color Brown or Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (C5H4N)SH
Quantity 10 g
Beilstein 21, II, 35
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,toluene,methanol and ethanol
IUPAC Name 1H-pyridine-4-thione
Formula Weight 111.16
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 4-Mercaptopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Mercaptopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.