missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptophenylacetic acid, 97%

Product Code. p-7030118
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486630 1 g 1g
11496630 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486630 Supplier Thermo Scientific Alfa Aesar Supplier No. H27658.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39161-84-7 | C8H8O2S | 168.21 g/mol

An useful synthetic intermediate for the preparation of novel anti inflammatory agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.

Solubility
Slightly soluble in water. Soluble in Dichloromethane, Ethanol and Methanol

Notes
Air sensitive. Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39161-84-7
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00797617
InChI Key ORXSLDYRYTVAPC-UHFFFAOYSA-N
Synonym 4-mercaptophenylacetic acid, 2-4-mercaptophenyl acetic acid, 2-4-sulfanylphenyl acetic acid, benzeneacetic acid, 4-mercapto, 4-mercapto-phenyl-acetic acid, acmc-20amfq, 4-mercaptobenzeneacetate, 4-mercapto-phenylacetic acid, 4-mercaptophenyl acetic acid, 4-mercapto-phenyl acetic acid
PubChem CID 3390511
IUPAC Name 2-(4-sulfanylphenyl)acetic acid
SMILES OC(=O)CC1=CC=C(S)C=C1

Specifications

Melting Point 105°C to 110°C
Quantity 1 g
Sensitivity Air sensitive
Solubility Information Slightly soluble in water. Soluble in Dichloromethane,Ethanol and Methanol
Formula Weight 168.21
Percent Purity 97%
Chemical Name or Material 4-Mercaptophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.