missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptophenol, TRC

Product Code. 30474043
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474043 Supplier Toronto Research Chemicals Leverantörsnummer TRCM2544201G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 4-Mercaptophenol
CAS 637-89-8
Molecular Formula C6H6OS
Quantity 1 g
Synonym p-Mercaptophenol, 4-Hydroxybenzenethiol, 4-Hydroxythiophenol, 4-Mercaptophenol, 4-Merpcatophenol, Monothiohydroquinone, NSC 46192, Thiohydroquinone, p-Hydroxybenzenethiol, p-Hydroxythiophenol, p-Mercaptophenol
InChI Formula InChI=1S/C6H6OS/c7-5-1-3-6(8)4-2-5/h1-4,7-8H
SMILES Oc1ccc(S)cc1
IUPAC Name 4-sulfanylphenol
Molecular Weight (g/mol) 126.18
Formula Weight 126.0139
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.