missing translation for 'onlineSavingsMsg'
Learn More

4-(Maleimidomethyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15489695
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315276-43-7
Molecular Formula C17H20BNO4
Molecular Weight (g/mol) 313.16
MDL Number MFCD18827482
InChI Key DROVAVNVMSFFHB-UHFFFAOYSA-N
Synonym 4-maleimidomethyl benzeneboronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrole-2,5-dione, amtb376, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl maleimide, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-1h-pyrrole-2,5-dione, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-pyrrole-2,5-dione
PubChem CID 71306538
IUPAC Name 1-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]pyrrole-2,5-dione
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3C(=O)C=CC3=O

Specifications

Quantity 1 g
Formula Weight 313.16
Percent Purity 95%
Chemical Name or Material 4-(Maleimidomethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'