missing translation for 'onlineSavingsMsg'
Learn More

4-Maleimidobutyric acid N-hydroxysuccinimide ester, 97%

Product Code. 10103434 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10103434 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10103434 Supplier Thermo Scientific Acros Supplier No. 398681000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 80307-12-6 | C12H12N2O6 | 280.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 80307-12-6
Melting Point 124.0°C to 132.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C12H12N2O6
MDL Number MFCD00036817
Quantity 100 mg
Synonym gmbs, 4-maleimidobutyric acid n-hydroxysuccinimide ester, 4-maleimidobutyric acid n-succinimidyl ester, n-gamma-maleimidobutyryloxy succinimide, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl butanoate, n-4-maleimidobutyryloxy succinimide, n-succinimidyl 4-maleimidobutyrate, n-succinimidyl-4-maleimidobutyrate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate
InChI Key PVGATNRYUYNBHO-UHFFFAOYSA-N
SMILES O=C(CCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-(2,5-dioxopyrrol-1-yl)butanoate
Molecular Weight (g/mol) 280.24
PubChem CID 133440
Formula Weight 280.23
Percent Purity 97%
Chemical Name or Material 4-Maleimidobutyric acid N-hydroxysuccinimide ester
Show More Show Less
compliance-icons
Product Identifier
  • 4-Maleimidobutyric acid N-hydroxysuccinimide ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.