missing translation for 'onlineSavingsMsg'
Learn More

4-L-Glutamic Acid Desmopressin TFA Salt, TRC

Product Code. 30389388
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389388 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389388 Supplier Toronto Research Chemicals Supplier No. TRCG29670525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-L-Glutamic Acid Desmopressin TFA Salt
Analyte or Component Names 4-L-Glutamic Acid Desmopressin TFA Salt
Shipping Condition Room Temperature
Molecular Formula C46H63N13O13S2 • (C2HF3O2)
Synonym Desmopresssin Impurity B
InChI Formula InChI=1S/C46H63N13O13S2.C2HF3O2/c47-35(61)22-32-43(70)58-33(45(72)59-18-5-9-34(59)44(71)55-28(8-4-17-51-46(49)50)39(66)52-23-36(48)62)24-74-73-19-16-37(63)53-30(21-26-10-12-27(60)13-11-26)41(68)56-31(20-25-6-2-1-3-7-25)42(69)54-29(40(67)57-32)14-15-38(64)65;3-2(4,5)1(6)7/h1-3,6-7,10-13,28-34,60H,4-5,8-9,14-24H2,(H2,47,61)(H2,48,62)(H,52,66)(H,53,63)(H,54,69)(H,55,71)(H,56,68)(H,57,67)(H,58,70)(H,64,65)(H4,49,50,51);(H,6,7)/t28-,29-,30-,31-,32-,33-,34-;/m0./s1
Solution Type Neat
SMILES O=C(N1CCC[C@H]1C(N[C@H](C(NCC(N)=O)=O)CCCNC(N)=N)=O)[C@@H](NC([C@H](CC(N)=O)NC([C@H](CCC(O)=O)NC([C@]([H])(CC2=CC=CC=C2)N3)=O)=O)=O)CSSCCC(N[C@@H](CC4=CC=C(C=C4)O)C3=O)=O.O=C(O)C(F)(F)F
IUPAC Name 2,2,2-trifluoroacetic acid--3-((4R,7S,10S,13S,16S)-7-(2-amino-2-oxoethyl)-4-((S)-2-(((S)-1-((2-amino-2-oxoethyl)amino)-5-guanidino-1-oxopentan-2-yl)carbamoyl)pyrrolidine-1-carbonyl)-13-benzyl-16-(4-hydroxybenzyl)-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloicosan-10-yl)propanoic acid (1/1)
Industry Type Pharmaceutical
Molecular Weight (g/mol) 1183.4
Quantity 25 mg
Formula Weight 1184.22
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.