missing translation for 'onlineSavingsMsg'
Learn More

4-Keto Retinal, TRC

Product Code. 30402998
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402998 10 mg -
30393374 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402998 Supplier Toronto Research Chemicals Supplier No. TRCK20495010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33532-44-4
Molecular Formula C20 H26 O2
Molecular Weight (g/mol) 298.42
Synonym Retinal, 4-oxo-, 4-Oxoretinal, 4-Ketoretinal, 4-Oxo-all-trans-retinal, 4-keto-Retinal, all-trans-4-Oxoretinal, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-3-oxo-cyclohexen-1-yl)nona-2,4,6,8-tetraenal
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)nona-2,4,6,8-tetraenal
SMILES C\C(=C/C=O)\C=C\C=C(/C)\C=C\C1=C(C)C(=O)CCC1(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H26O2/c1-15(7-6-8-16(2)12-14-21)9-10-18-17(3)19(22)11-13-20(18,4)5/h6-10,12,14H,11,13H2,1-5H3/b8-6+,10-9+,15-7+,16-12+
Formula Weight 298.1933
Chemical Name or Material 4-Ketoretinal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.