missing translation for 'onlineSavingsMsg'
Learn More

4' '-Keto-5-O-[(2-propenyloxy)carbonyl]-Abamectin, TRC

Product Code. 30395097
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395097 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395097 Supplier Toronto Research Chemicals Supplier No. TRCK816820100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4''-Keto-5-O-[(2-propenyloxy)carbonyl]-Abamectin
Molecular Formula C52 H74 O16
Quantity 100 mg
InChI Formula InChI=1S/C52H74O16/c1-12-21-59-50(55)66-46-32(7)22-38-49(54)63-37-23-36(67-51(26-37)20-19-31(6)45(68-51)28(3)13-2)18-17-30(5)44(29(4)15-14-16-35-27-60-48(46)52(35,38)56)64-42-25-40(58-11)47(34(9)62-42)65-41-24-39(57-10)43(53)33(8)61-41/h12,14-17,19-20,22,28-29,31,33-34,36-42,44-48,56H,1,13,18,21,23-27H2,2-11H3/b15-14+,30-17+,35-16+/t28-,29-,31-,33-,34-,36+,37-,38-,39-,40-,41-,42-,44-,45+,46+,47-,48+,51+,52+/m0/s1
SMILES CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)C(=O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](OC(=O)OCC=C)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C
IUPAC Name [(1'R,2R,3S,4'S,6S,8'R,10'E,12'S,13'S,14'E,16'E,20'R,21'R,24'S)-2-[(2S)-butan-2-yl]-24'-hydroxy-12'-[(2R,4S,5S,6S)-4-methoxy-5-[(2R,4S,6S)-4-methoxy-6-methyl-5-oxooxan-2-yl]oxy-6-methyloxan-2-yl]oxy-3,11',13',22'-tetramethyl-2'-oxospiro[2,3-dihydropyran-6,6'-3,7,19-trioxatetracyclo[15.6.1.14,8.020,24]pentacosa-10,14,16,22-tetraene]-21'-yl] prop-2-enyl carbonate
Molecular Weight (g/mol) 955.134
Formula Weight 954.498
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.