missing translation for 'onlineSavingsMsg'
Learn More

4-Keto 13-cis-Retinoic Acid-d3, TRC

Product Code. 30389396
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389396 0.5 mg -
30399545 1 mg 1mg
30402081 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30389396 Supplier Toronto Research Chemicals Supplier No. TRCK2049720.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185241-31-9
Molecular Formula C20H23D3O3
Molecular Weight (g/mol) 317.44
Synonym 3,7-Dimethyl-9-(4-keto-2,6,6-trimethyl-1-cyclohexen-1-yl)-2Z,4E,6E,8E-nonatetranenoic Acid-d3, 4-Oxoisotretinoin-d3, Ro 22-6595-d3, 4-Oxoretinoic Acid-d3
IUPAC Name 9-[6,6-dimethyl-3-oxo-2-(trideuteriomethyl)cyclohexen-1-yl]-3,7-dimethylnona-2,4,6,8-tetraenoic acid
SMILES O=C1CCC(C)(C)C(/C=C/C(C)=C/C=C/C(C)=C\C(O)=O)=C1C([2H])([2H])[2H]

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C20H26O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13H,11-12H2,1-5H3,(H,22,23)/i3D3
Formula Weight 317.207
Chemical Name or Material 4-Keto 13-cis-Retinoic Acid-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.