missing translation for 'onlineSavingsMsg'
Learn More

4-Isothiocyanatophenyl Alpha-Nigeroside, TRC

Product Code. 30397149
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397149 0.5 mg 0.5mg
30404603 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397149 Supplier Toronto Research Chemicals Supplier No. TRCI9037000.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Isothiocyanatophenyl alpha-Nigeroside
Analyte or Component Names 4-Isothiocyanatophenyl alpha-Nigeroside
Shipping Condition Room Temperature
Molecular Formula C19H25NO11S
Synonym 4-Isothiocyanatophenyl 3-O-α-D-Glucopyranosyl-α-D-glucopyranoside
InChI Formula InChI=1S/C19H25NO11S/c21-5-10-12(23)14(25)15(26)18(29-10)31-17-13(24)11(6-22)30-19(16(17)27)28-9-3-1-8(2-4-9)20-7-32/h1-4,10-19,21-27H,5-6H2/t10-,11-,12-,13-,14+,15-,16-,17+,18-,19+/m1/s1
Solution Type Neat
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](CO)O[C@H](OC3=CC=C(N=C=S)C=C3)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3R,4S,5S,6R)-2-(((2R,3R,4S,5R,6R)-3,5-dihydroxy-2-(hydroxymethyl)-6-(4-isothiocyanatophenoxy)tetrahydro-2H-pyran-4-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 475.47
Quantity 0.5 mg
Formula Weight 475.11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.