missing translation for 'onlineSavingsMsg'
Learn More

4-isopropylphenyl isothiocyanate, Thermo Scientific™

Product Code. 10459454
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10459454 5 g 5g
10344682 1 g 1g
10704773 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10459454 Supplier Thermo Scientific Maybridge Supplier No. SB01635DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89007-45-4
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.27
MDL Number MFCD00022060
InChI Key SNWJJTIJIDXIHL-UHFFFAOYSA-N
Synonym 4-isopropylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanatobenzene, 4-isopropylphenylisothiocyanate, 1-isothiocyanato-4-propan-2-yl benzene, 4-methylethyl benzenisothiocyanate, acmc-20amft, 4isopropylphenylisothiocyanate, 4-iso-propylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanato-benzene, 1-isopropyl-4-isothiocyanatobenzene #
PubChem CID 145194
IUPAC Name 1-isothiocyanato-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)N=C=S

Specifications

Density 1.0300g/mL
Assay Percent Range 95%+
Linear Formula (CH3)2CHC6H4NCS
Quantity 5 g
Formula Weight 177.27
Percent Purity 95%
Chemical Name or Material 4-Isopropylphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.