missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenyl isocyanate, 98%

Product Code. 11367636
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367636 10 g 10g
11357636 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367636 Supplier Thermo Scientific Alfa Aesar Supplier No. L11217.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 31027-31-3 | C10H11NO | 161.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31027-31-3
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00013880
InChI Key PJVRNNRZWASOIT-UHFFFAOYSA-N
Synonym 4-isopropylphenyl isocyanate, 4-isopropylphenylisocyanate, 1-isocyanato-4-isopropylbenzene, 1-isocyanato-4-isopropyl-benzene, p-isopropylphenyl isocyanate, benzene, 1-isocyanato-4-1-methylethyl, 4-isopropyl phenylisocyanate, 4-methylethyl benzenisocyanate, p-cumenyl isocyanate, acmc-1aild
PubChem CID 94416
IUPAC Name 1-isocyanato-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)N=C=O

Specifications

Density 1.01
Boiling Point 84°C to 85°C (4 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.5205
Quantity 10 g
UN Number UN2206
Beilstein 2936403
Sensitivity Moisture sensitive
Formula Weight 161.21
Percent Purity 98%
Chemical Name or Material 4-Isopropylphenyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Isopropylphenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: p-Isopropylphenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.