missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenyl isocyanate, 98%, ACROS Organics™

Product Code. 10083730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10083730 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083730 Supplier Thermo Scientific Acros Supplier No. 314090050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 31027-31-3
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or Orange
Density 1.0100g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H11NO
Linear Formula (CH3)2CHC6H4NCO
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)
MDL Number MFCD00013880
Quantity 5 g
Synonym 4-isopropylphenyl isocyanate, 4-isopropylphenylisocyanate, 1-isocyanato-4-isopropylbenzene, 1-isocyanato-4-isopropyl-benzene, p-isopropylphenyl isocyanate, benzene, 1-isocyanato-4-1-methylethyl, 4-isopropyl phenylisocyanate, 4-methylethyl benzenisocyanate, p-cumenyl isocyanate, acmc-1aild
InChI Key PJVRNNRZWASOIT-UHFFFAOYSA-N
SMILES CC(C)C1=CC=C(C=C1)N=C=O
IUPAC Name 1-isocyanato-4-propan-2-ylbenzene
Molecular Weight (g/mol) 161.204
PubChem CID 94416
Formula Weight 161.21
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 4-Isopropylphenyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.