missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenol, TRC

Product Code. 30402675
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402675 50 g 50g
30394563 250 g -
30392329 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402675 Supplier Toronto Research Chemicals Supplier No. TRCI87271050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-89-8
Molecular Formula C9 H12 O
Molecular Weight (g/mol) 136.19
Synonym 4-(1-Methylethyl)phenol, Propofol Imp. H (EP), Phenol, 4-(1-methylethyl)- (9CI, ACI), Phenol, p-isopropyl- (8CI), 4-(Propan-2-yl)phenol, 4-Isopropylphenol, Australol, NSC 1888, p-Cumenol, p-Isopropylphenol
IUPAC Name 4-propan-2-ylphenol
SMILES CC(C)c1ccc(O)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H12O/c1-7(2)8-3-5-9(10)6-4-8/h3-7,10H,1-2H3
Formula Weight 136.0888
Chemical Name or Material 4-Isopropylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.