missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenol, TRC

Product Code. 30394563
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394563 250 g -
30402675 50 g 50g
30392329 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394563 Supplier Toronto Research Chemicals Supplier No. TRCI872710250G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 99-89-8
Molecular Formula C9 H12 O
Molecular Weight (g/mol) 136.19
Synonym 4-(1-Methylethyl)phenol, Propofol Imp. H (EP), Phenol, 4-(1-methylethyl)- (9CI, ACI), Phenol, p-isopropyl- (8CI), 4-(Propan-2-yl)phenol, 4-Isopropylphenol, Australol, NSC 1888, p-Cumenol, p-Isopropylphenol
IUPAC Name 4-propan-2-ylphenol
SMILES CC(C)c1ccc(O)cc1

Specifica

Quantity 250 g
InChI Formula InChI=1S/C9H12O/c1-7(2)8-3-5-9(10)6-4-8/h3-7,10H,1-2H3
Formula Weight 136.0888
Chemical Name or Material 4-Isopropylphenol
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.