missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzyl alcohol, 97%

Product Code. 10103970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103970 25 g Glass bottle 25g
10447570 100 g Glass bottle 100g
10657114 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103970 Supplier Thermo Scientific Acros Supplier No. 187800250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 536-60-7 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-60-7
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00004663
InChI Key OIGWAXDAPKFNCQ-UHFFFAOYSA-N
Synonym 4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol
PubChem CID 325
ChEBI CHEBI:27628
IUPAC Name (4-propan-2-ylphenyl)methanol
SMILES CC(C)C1=CC=C(C=C1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 28°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 248°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHC6H4CH2OH
Refractive Index 1.5180 to 1.52
Quantity 25 g
Beilstein 06, III, 1911
Merck Index 15, 2609
Specific Gravity 0.98
Formula Weight 150.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Isopropylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.