missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzeneboronic acid, 98+%

Product Code. 11318436
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318436 1 g 1g
11328436 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318436 Supplier Thermo Scientific Alfa Aesar Supplier No. L17459.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16152-51-5 | C9H13BO2 | 164.011 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16152-51-5
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.011
MDL Number MFCD01074614
InChI Key IAEUFBDMVKQCLU-UHFFFAOYSA-N
Synonym 4-isopropylphenylboronic acid, 4-isopropylbenzeneboronic acid, 4-isopropylphenyl boronic acid, 4-propan-2-ylphenyl boronic acid, p-isopropylphenylboronic acid, p-cumylboronic acid, 4-cumylboronic acid, 4-iso-propylphenyl boronic acid, 4-iso-propylphenylboronic acid
PubChem CID 2773480
IUPAC Name (4-propan-2-ylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C(C)C)(O)O

Specifications

Melting Point 142°C to 144°C
Quantity 1 g
Beilstein 2830262
Formula Weight 164.01
Percent Purity ≥98%
Chemical Name or Material 4-Isopropylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.