missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzaldehyde, 98%

Product Code. 10603522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10603522 5 g Glass bottle 5g
10376111 100 g Glass bottle 100g
10595861 500 g Glass bottle 500g
10686174 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10603522 Supplier Thermo Scientific Acros Supplier No. 334880050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-03-2 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-03-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00006953
InChI Key WTWBUQJHJGUZCY-UHFFFAOYSA-N
Synonym 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
PubChem CID 326
ChEBI CHEBI:28671
IUPAC Name 4-propan-2-ylbenzaldehyde
SMILES CC(C)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9790g/mL
Boiling Point 234°C to 235°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5290 to 1.532
Quantity 5 g
Merck Index 15, 2611
Specific Gravity 0.979
Solubility Information Solubility in water: insoluble
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Isopropylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.