missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylaniline, 99%

Product Code. 10216090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216090 50 g Glass bottle 50g
10601915 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216090 Supplier Thermo Scientific Acros Supplier No. 174530500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-88-7 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-88-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007900
InChI Key LRTFPLFDLJYEKT-UHFFFAOYSA-N
Synonym 4-isopropylaniline, cumidine, 4-aminocumene, p-isopropylaniline, p-cumidine, 4-propan-2-yl aniline, para-isopropylaniline, 4-amino-1-isopropylbenzene, cumene, p-amino, aniline, p-isopropyl
PubChem CID 7464
ChEBI CHEBI:43405
IUPAC Name 4-propan-2-ylaniline
SMILES CC(C)C1=CC=C(C=C1)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -63°C
Color Brown to Brown-Red
Density 0.9890g/mL
Boiling Point 226°C to 228°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHC6H4NH2
Refractive Index 1.5420 to 1.544
Quantity 50 g
Beilstein 12, 1147
Merck Index 15, 2610
Specific Gravity 0.989
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols
Formula Weight 135.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Isopropylaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Isopropylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.