missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylaniline, 97%, Maybridge

Product Code. 10634395
Change view
Click to view available options
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10634395 Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10634395 Supplier Thermo Scientific Maybridge Supplier No. SB00317EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 99-88-7
CAS Min % 97.0
Density 0.9890g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C9H13N
Linear Formula (CH3)2CHC6H4NH2
MDL Number MFCD00007900
Quantity 10 g
Synonym 4-isopropylaniline, cumidine, 4-aminocumene, p-isopropylaniline, p-cumidine, 4-propan-2-yl aniline, para-isopropylaniline, 4-amino-1-isopropylbenzene, cumene, p-amino, aniline, p-isopropyl
InChI Key LRTFPLFDLJYEKT-UHFFFAOYSA-N
SMILES CC(C)C1=CC=C(C=C1)N
IUPAC Name 4-propan-2-ylaniline
Molecular Weight (g/mol) 135.21
PubChem CID 7464
ChEBI CHEBI:43405
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 4-Isopropylaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.