missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropyl-3-methylaniline hydrochloride, 97%, Thermo Scientific™

Product Code. 10215314
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10215314 25 g 25g
10304732 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215314 Supplier Thermo Scientific Maybridge Supplier No. SB01749EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4534-11-6
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00156562
InChI Key VPOAQDSAPBSOLW-UHFFFAOYSA-N
ChEBI CHEBI:51926

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 25 g
Synonym ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester
SMILES [H+].[Cl-].CC(C)C1=CC=C(N)C=C1C
IUPAC Name hydrogen 3-methyl-4-(propan-2-yl)aniline chloride
PubChem CID 95298
Formula Weight 185.7
Percent Purity 95%
Chemical Name or Material 4-Isopropyl-3-methylaniline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.