missing translation for 'onlineSavingsMsg'
Learn More

(4-Isopropoxyphenyl)hydrazine Hydrochloride, TRC

Product Code. 30394306
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394306 1 g 1g
30402092 500 mg -
30391821 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394306 Supplier Toronto Research Chemicals Supplier No. TRCI9183231G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104033-62-7
Molecular Formula C9H14N2O . HCl
Molecular Weight (g/mol) 202.68
Synonym N-(2,5-Diethoxyphenyl)benzamide, 2',5'-Diethoxybenzanilide, NSC 163154
IUPAC Name (4-isopropoxyphenyl)hydrazine hydrochloride
SMILES CC(C)OC1=CC=C(C=C1)NN.Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H14N2O.ClH/c1-7(2)12-9-5-3-8(11-10)4-6-9;/h3-7,11H,10H2,1-2H3;1H
Formula Weight 202.09
Chemical Name or Material (4-Isopropoxyphenyl)hydrazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.