missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxybenzaldehyde, 97%

Product Code. 11455093
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455093 5 g 5g
11445093 1 g 1g
11465093 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455093 Supplier Thermo Scientific Alfa Aesar Supplier No. A12760.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18962-05-5 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18962-05-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00052357
InChI Key WDANSDASCKBVKH-UHFFFAOYSA-N
Synonym 4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde
PubChem CID 250077
IUPAC Name 4-propan-2-yloxybenzaldehyde
SMILES CC(C)OC1=CC=C(C=C1)C=O

Specifications

Boiling Point 108°C to 110°C (5 mmHg)
Refractive Index 1.547
Quantity 5 g
Beilstein 742921
Sensitivity Air Sensitive
Formula Weight 164.21
Percent Purity 97%
Chemical Name or Material 4-Isopropoxybenzaldehyde
compliance-icons
Product Identifier
  • 4-Isopropoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.