missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Isopropoxy-2-methylbenzeneboronic acid, 98%
Click to view available options
Quantity:
1 g
Specifications
Melting Point | 112°C to 116°C |
Quantity | 1 g |
Formula Weight | 194.04 |
Percent Purity | 98% |
Chemical Name or Material | 4-Isopropoxy-2-methylbenzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'