missing translation for 'onlineSavingsMsg'
Learn More

4-(Isocyanatomethyl)benzonitrile, TRC

Product Code. 30402187
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402187 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402187 Supplier Toronto Research Chemicals Supplier No. TRCI7195601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1205556-81-5
Molecular Formula C9H6N2O
Quantity 1 g
Synonym 4-(Isocyanatomethyl)-benzonitrile
InChI Formula InChI=1S/C9H6N2O/c10-5-8-1-3-9(4-2-8)6-11-7-12/h1-4H,6H2
SMILES O=C=NCc1ccc(cc1)[N+]#[C-]
Molecular Weight (g/mol) 158.16
Formula Weight 158.048
Chemical Name or Material 4-(Isocyanatomethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.