missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isobutyramidobenzeneboronic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.
Solubility
Slightly Soluble (3.3 g/L) (25°C)
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.
Solubility
Slightly Soluble (3.3 g/L) (25°C)
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications
Melting Point | 235°C |
Quantity | 250 mg |
Solubility Information | Slightly Soluble (3.3g/L) (25°C) |
Formula Weight | 207.04 |
Percent Purity | 98% |
Chemical Name or Material | 4-Isobutyramidobenzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'