missing translation for 'onlineSavingsMsg'
Learn More

4-Isobutyramidobenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15489907
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489907 250 mg 250mg
15479907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489907 Supplier Thermo Scientific Alfa Aesar Supplier No. H53147.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Solubility
Slightly Soluble (3.3 g/L) (25°C)

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874219-50-8
Molecular Formula C10H14BNO3
Molecular Weight (g/mol) 207.036
MDL Number MFCD08235068
InChI Key DBWRRIVRDITVFV-UHFFFAOYSA-N
Synonym 4-isobutyramidobenzeneboronic acid, 4-isobutyramidophenyl boronic acid, 4-isobutyramidophenylboronic acid, 4-isobutyramido benzeneboronic acid, boronic acid,b-4-2-methyl-1-oxopropyl amino phenyl, 4-2-methylpropanamido phenylboronic acid, acmc-209qle, 4-isobutyramidophenyl boronicacid, 4-2-methylpropanamido phenyl boronic acid, 4-2-methylpropanoylamino phenyl boronic acid
PubChem CID 44119813
IUPAC Name [4-(2-methylpropanoylamino)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)NC(=O)C(C)C)(O)O

Specifications

Melting Point 235°C
Quantity 250 mg
Solubility Information Slightly Soluble (3.3g/L) (25°C)
Formula Weight 207.04
Percent Purity 98%
Chemical Name or Material 4-Isobutyramidobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.