Learn More
4'-Isobutylacetophenone, 97%
Beskrivelse
4'-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4′-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.
Solubility
Miscible with chloroform and methanol. Slightly miscible with water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with reducing agents, strong oxidizing agents and strong bases.
Tekniske data
Tekniske data
| Density | 0.952 |
| Flash Point | 54°C (129°F) |
| Refractive Index | 1.518 |
| Quantity | 250 g |
| UN Number | UN1224 |
| Beilstein | 1935275 |
| Solubility Information | Miscible with chloroform and methanol. Slightly miscible with water. |
| Formula Weight | 176.27 |
| Percent Purity | 97% |
| Chemical Name or Material | 4'-Isobutylacetophenone |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.