missing translation for 'onlineSavingsMsg'
Learn More

4'-Isobutyl-2,2-dibromopropiophenone, TRC

Product Code. 30389562
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389562 25 mg 25mg
30395223 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389562 Supplier Toronto Research Chemicals Supplier No. TRCI78040025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Isobutyl-2,2-dibromopropiophenone
CAS 104483-05-8
Molecular Formula C13H16Br2O
Quantity 25 mg
Synonym 2,2-Dibromo-1-[4-(2-methylpropyl)phenyl]-1-propanone
InChI Formula InChI=1S/C13H16Br2O/c1-9(2)8-10-4-6-11(7-5-10)12(16)13(3,14)15/h4-7,9H,8H2,1-3H3
SMILES CC(C)Cc1ccc(cc1)C(=O)C(C)(Br)Br
IUPAC Name 2,2-dibromo-1-[4-(2-methylpropyl)phenyl]propan-1-one
Molecular Weight (g/mol) 348.07
Formula Weight 345.9568
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.