missing translation for 'onlineSavingsMsg'
Learn More

4-Iodotoluene, 98%

Product Code. 11417527
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417527 25 g 25g
11427527 100 g 100g
11437527 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417527 Supplier Thermo Scientific Alfa Aesar Supplier No. A15631.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-31-7 | C7H7I | 218.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Iodotoluene is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals.

Solubility
Solubility in methanol is almost transparent.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-31-7
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001059
InChI Key UDHAWRUAECEBHC-UHFFFAOYSA-N
Synonym 4-iodotoluene, p-iodotoluene, benzene, 1-iodo-4-methyl, toluene, p-iodo, p-tolyl iodide, p-methyliodobenzene, 1-methyl-4-iodobenzene, 1-iodo-4-methyl-benzene, unii-g75kcn1am8, iodotoluene 4-
PubChem CID 12207
IUPAC Name 1-iodo-4-methylbenzene
SMILES CC1=CC=C(C=C1)I

Specifications

Melting Point 33°C to 35°C
Density 1.678
Boiling Point 210°C to 212°C
Flash Point 90°C (194°F)
Quantity 25 g
Beilstein 1903637
Sensitivity Light sensitive
Formula Weight 218.04
Percent Purity 98%
Chemical Name or Material 4-Iodotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.