missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenylacetonitrile, 97%

Product Code. 11398146
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398146 1 g 1g
11308156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398146 Supplier Thermo Scientific Alfa Aesar Supplier No. L15119.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51628-12-7 | C8H6IN | 243.047 g/mol

4-Iodophenylacetonitrile is used as an intermediate in organic syntheses and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodophenylacetonitrile is used as an intermediate in organic syntheses and in pharmaceuticals.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51628-12-7
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.047
MDL Number MFCD00060306
InChI Key PNXWQTYSBFGIFD-UHFFFAOYSA-N
Synonym 4-iodophenylacetonitrile, 2-4-iodophenyl acetonitrile, 4-iodobenzyl cyanide, 4-iodophenyl acetonitrile, benzeneacetonitrile, 4-iodo, p-iodobenzylcyanide, pubchem19992, acmc-1asuk, 4-cyanomethyl phenyl iodide, 4-iodo-phenyl-acetonitrile
PubChem CID 142866
IUPAC Name 2-(4-iodophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)I

Specifications

Melting Point 52°C to 54°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3439
Beilstein 1934479
Sensitivity Light sensitive
Solubility Information Insoluble in water
Formula Weight 243.05
Percent Purity 97%
Chemical Name or Material 4-Iodophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.