missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenyl isocyanate, 97%

Product Code. 11342847
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11342847

Brand: Thermo Scientific Alfa Aesar L16000.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15845-62-2 | C7H4INO | 245.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15845-62-2
Molecular Formula C7H4INO
Molecular Weight (g/mol) 245.019
MDL Number MFCD00013875
InChI Key KVMKUTIUJGYHBA-UHFFFAOYSA-N
Synonym 4-iodophenyl isocyanate, 4-iodophenylisocyanate, 1-iodo-4-isocyanato-benzene, p-iodophenylisocyanate, acmc-1bsbj, benzene,1-iodo-4-isocyanato
PubChem CID 3399787
IUPAC Name 1-iodo-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)I

Specifications

Melting Point 43°C to 47°C
Boiling Point 84°C (5 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 1 g
UN Number UN2811
Beilstein 2716151
Sensitivity Light and moisture sensitive
Formula Weight 245.02
Percent Purity 97%
Chemical Name or Material 4-Iodophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.