missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzylamine hydrochloride, 97%

Product Code. 11334356
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334356 5 g 5g
11324356 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334356 Supplier Thermo Scientific Alfa Aesar Supplier No. L19967.06
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 59528-27-7 | C7H9ClIN | 269.51 g/mol

4-Iodobenzylamine hydrochloride is used to prepare cyclic amide by reacting with methyl 4-bromomethyl-3-methoxycarbonylcinnamate in the presence of triethylamine. It is also used in pharmaceuticals, agrochemicals and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzylamine hydrochloride is used to prepare cyclic amide by reacting with methyl 4-bromomethyl-3-methoxycarbonylcinnamate in the presence of triethylamine. It is also used in pharmaceuticals, agrochemicals and dye industries.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59528-27-7
Molecular Formula C7H9ClIN
Molecular Weight (g/mol) 269.51
MDL Number MFCD04039324
InChI Key GBJMURRFWZREHE-UHFFFAOYSA-N
Synonym 4-iodobenzylamine hydrochloride, 4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine hcl, benzenemethanamine, 4-iodo-, hydrochloride, 4-iodophenyl methylamine, chloride, 1-4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine, hydrochloride salt, 4-iodobenzylaminehydrochloride, pubchem3304, acmc-209tzk
PubChem CID 16217505
IUPAC Name (4-iodophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC=C1CN)I.Cl

Specifications

Melting Point 275°C to 280°C (decomposition)
Quantity 5 g
Beilstein 3691299
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 269.51
Percent Purity 97%
Chemical Name or Material 4-Iodobenzylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.