missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl bromide, 97%

Product Code. 11914531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11914531 25 g Glass bottle 25g
10294841 1 g Glass bottle 1g
10366641 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11914531 Supplier Thermo Scientific Acros Supplier No. 342460250
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16004-15-2 | C7H6BrI | 296.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16004-15-2
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.93
InChI Key BQTRMYJYYNQQGK-UHFFFAOYSA-N
Synonym 4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide
PubChem CID 954258
IUPAC Name 1-(bromomethyl)-4-iodobenzene
SMILES C1=CC(=CC=C1CBr)I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 77.0°C to 82.0°C
Color Yellow
Boiling Point 140.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4CH2Br
Quantity 25 g
Formula Weight 296.93
Percent Purity 97%
Physical Form Adhering Crystalline Powder
Chemical Name or Material 4-Iodobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.