missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl bromide, 97%

Product Code. 11483727
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483727 25 g 25g
11473727 5 g 5g
11493727 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483727 Supplier Thermo Scientific Alfa Aesar Supplier No. A18200.14
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16004-15-2 | C7H6BrI | 296.933 g/mol

4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, bases and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16004-15-2
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
MDL Number MFCD00209656
InChI Key BQTRMYJYYNQQGK-UHFFFAOYSA-N
Synonym 4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide
PubChem CID 954258
IUPAC Name 1-(bromomethyl)-4-iodobenzene
SMILES C1=CC(=CC=C1CBr)I

Specifications

Melting Point 78°C to 82°C
Boiling Point 140°C (15 mmHg)
Odor Irritating
Quantity 25 g
UN Number UN3261
Beilstein 2325160
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 296.94
Percent Purity 97%
Chemical Name or Material 4-Iodobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1-Bromomethyl-4-iodebenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.