missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzenesulfonamide, 95%

Product Code. 15355038 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15355038 1 g 1g
15365038 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15355038 Supplier Thermo Scientific Acros Supplier No. 455700010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 825-86-5 | C6H6INO2S | 283.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 825-86-5
Molecular Formula C6H6INO2S
Molecular Weight (g/mol) 283.09
InChI Key KQZFABSTXSNEQH-UHFFFAOYSA-N
Synonym p-iodobenzenesulfonamide, 4-iodobenzenesulphonamide, 4-iodobenzene-1-sulfonamide, benzenesulfonamide,4-iodo, benzenesulfonamide, p-iodo, p-jodbenzolsulfonamid, 4-iodibenzenesulphonamide, 4-iodo-benzenesulfonamide, 4-iodo-1-sulfamoylbenzene, 3-11-00-00110 beilstein handbook reference
PubChem CID 13218
IUPAC Name 4-iodobenzenesulfonamide
SMILES C1=CC(=CC=C1S(=O)(=O)N)I

Specifications

Melting Point 193.0°C to 195.0°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Linear Formula IC6H4SO2NH2
Quantity 1 g
Formula Weight 283.09
Percent Purity 95%
Chemical Name or Material 4-Iodobenzenesulfonamide
compliance-icons
Product Identifier
  • 4-Iodobenzenesulfonamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.