missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzaldehyde, 98+%

Product Code. 11378416
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378416 250 mg 250mg
11358416 1 g 1g
11368416 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378416 Supplier Thermo Scientific Alfa Aesar Supplier No. L17448.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15164-44-0 | C7H5IO | 232.02 g/mol

4-Iodobenzaldehyde is used in synthesis of 4-[2-(trimethylsilyl)ethynyl]benzaldehyde, 5,15-dimesityl-10-(3-[2-(trimethylsilyl)ethynyi]phenyl}-20-(4-iodophenyl)porphyrin, and 5,15-dimesityl-10-[3,5-bis{2-[4-(N,N?-difluoroboryl-1,9-dimethyidipyrrin-5-yl)-phenyl]ethynyl}phenyl]-20-(4-iodophenyl)porphyrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzaldehyde is used in synthesis of 4-[2-(trimethylsilyl)ethynyl]benzaldehyde, 5,15-dimesityl-10-(3-[2-(trimethylsilyl)ethynyi]phenyl}-20-(4-iodophenyl)porphyrin, and 5,15-dimesityl-10-[3,5-bis{2-[4-(N,N′-difluoroboryl-1,9-dimethyidipyrrin-5-yl)-phenyl]ethynyl}phenyl]-20-(4-iodophenyl)porphyrin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air and light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15164-44-0
Molecular Formula C7H5IO
Molecular Weight (g/mol) 232.02
MDL Number MFCD00039576
InChI Key NIEBHDXUIJSHSL-UHFFFAOYSA-N
Synonym p-iodobenzaldehyde, benzaldehyde, 4-iodo, 4-iodo-benzaldehyde, iodobenzenecarbaldehyde, 4-iodo benzaldehyde, pubchem5349, acmc-1bpp9, intermediates-zcf02160, 4-iodobenzaldehyde, ksc181a1r
PubChem CID 96657
IUPAC Name 4-iodobenzaldehyde
SMILES IC1=CC=C(C=O)C=C1

Specifications

Melting Point 76°C to 78°C
Quantity 250 mg
Beilstein 636101
Sensitivity Air and light sensitive
Solubility Information Insoluble in water.
Formula Weight 232.02
Percent Purity ≥98%
Chemical Name or Material 4-Iodobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.