missing translation for 'onlineSavingsMsg'
Learn More

4-Iodoanisole, 98+%

Product Code. 11470113
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470113 100 g 100g
11460113 25 g 25g
11480113 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470113 Supplier Thermo Scientific Alfa Aesar Supplier No. A12144.22
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 696-62-8 | C7H7IO | 234.036 g/mol

4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products, pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.

Solubility
Soluble in ethanol, ether, chloroform.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-62-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001056
InChI Key SYSZENVIJHPFNL-UHFFFAOYSA-N
Synonym 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene
PubChem CID 69676
IUPAC Name 1-iodo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)I

Specifications

Melting Point 48°C to 52°C
Boiling Point 240°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 1906692
Sensitivity Light sensitive
Solubility Information Soluble in ethanol,ether,chloroform.
Formula Weight 234.04
Percent Purity ≥98%
Chemical Name or Material 4-Iodoanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.