missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodoanisole, 98+%, Thermo Scientific Chemicals
19.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products, pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.
Solubility
Soluble in ethanol, ether, chloroform.
Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.
Solubility
Soluble in ethanol, ether, chloroform.
Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
Specifications
Melting Point | 48°C to 52°C |
Boiling Point | 240°C |
Flash Point | >110°C (230°F) |
Quantity | 25 g |
Beilstein | 1906692 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in ethanol,ether,chloroform. |
Formula Weight | 234.04 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Iodoanisole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'