missing translation for 'onlineSavingsMsg'
Learn More

4-Iodoaniline, 98%

Product Code. 11495027
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495027 250 g 250g
11485027 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495027 Supplier Thermo Scientific Alfa Aesar Supplier No. B21936.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 540-37-4 | C6H6IN | 219.025 g/mol

4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).

Solubility
Slightly soluble in water.Soluble in chloroform, methanol.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-37-4
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.025
MDL Number MFCD00007848
InChI Key VLVCDUSVTXIWGW-UHFFFAOYSA-N
Synonym p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
PubChem CID 10893
IUPAC Name 4-iodoaniline
SMILES C1=CC(=CC=C1N)I

Specifications

Melting Point 61°C to 64°C
Quantity 250 g
Beilstein 774101
Sensitivity Light sensitive
Merck Index 14,5027
Solubility Information Slightly soluble in water.Soluble in chloroform,methanol.
Formula Weight 219.03
Percent Purity 98%
Chemical Name or Material 4-Iodoaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.