missing translation for 'onlineSavingsMsg'
Learn More

4'-Iodoacetanilide, 97%

Product Code. 10267873 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10267873 1 g Glass bottle 1g
10084313 5 g Glass bottle 5g
10146163 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10267873 Supplier Thermo Scientific Acros Supplier No. 393120010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-50-4 | C8H8INO | 261.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-50-4
Molecular Formula C8H8INO
Molecular Weight (g/mol) 261.06
MDL Number MFCD00016352
InChI Key SIULLDWIXYYVCU-UHFFFAOYSA-N
Synonym n-4-iodophenyl acetamide, 4-iodoacetanilide, 4'-iodoacetanilide, p-iodoacetanilide, acetamide, n-4-iodophenyl, acetanilide, 4'-iodo, 4'-iodo-acetanilide, pubchem3293, acmc-20akb8, maybridge1_006480
PubChem CID 12147
IUPAC Name N-(4-iodophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 184.0°C to 185.0°C
Color Blue-Gray to Purple
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula IC6H4NHC(=O)CH3
Quantity 1 g
Formula Weight 261.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4'-Iodoacetanilide
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Iodoacetanilide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.