missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-m-xylene, 98%

Product Code. 11482908
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482908 50 g 50g
11472908 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482908 Supplier Thermo Scientific Alfa Aesar Supplier No. A16878.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4214-28-2 | C8H9I | 232.06 g/mol

4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4214-28-2
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00013706
InChI Key BUNKQJAMHYKQIM-UHFFFAOYSA-N
Synonym 4-iodo-m-xylene, 2,4-dimethyliodobenzene, 1,3-dimethyl-4-iodobenzene, m-xylene, 4-iodo, benzene, 1-iodo-2,4-dimethyl, 2,4-dimethyl-1-iodobenzene, 2-iodo-1,5-dimethylbenzene, 4-jod-m-xylol, pubchem3869, acmc-209jnr
PubChem CID 77885
IUPAC Name 1-iodo-2,4-dimethylbenzene
SMILES CC1=CC=C(I)C(C)=C1

Specifications

Density 1.628
Quantity 50 g
Refractive Index 1.598
Beilstein 1422602
Boiling Point 231°C to 232°C
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 4-Iodo-m-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.